Gentamicin ELISA Kit - Extracellular
Lieferant: Abcam
Gentamicin ELISA kit - Extracellular is a single-wash 90-min Simplestep used to quantify Gentamicin with a sensitivity of 55,11 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Competitive ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders.