Mouse Lipocalin-2 ELISA Kit (NGAL)
Lieferant: Abcam
AB199083-96 AB199083-10X96
ABCAAB199083-96EA 975 EUR
ABCAAB199083-96 ABCAAB199083-10X96
Mouse Lipocalin-2 ELISA Kit (NGAL)
Tests ELISAs
Mouse Lipocalin-2 ELISA kit (NGAL) is a single-wash 90-min Simplestep used to quantify Mouse Lipocalin-2 (NGAL) with a sensitivity of 1,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 15 citations.
Zertifizierungen: RUO