Anti-Cyclin D2 Rabbit Monoclonal Antibody [clone: EPR27468-62]
Lieferant: Abcam
AB308258-100 AB308258-20UL AB308258-1ML
ABCAAB308258-100EA 694 EUR
ABCAAB308258-100 ABCAAB308258-20UL ABCAAB308258-1ML
Anti-Cyclin D2 Rabbit Monoclonal Antibody [clone: EPR27468-62]
Antikörper
Rabbit monoclonal [EPR27468-62] to Cyclin D2.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Cyclin D2
Clonality: Monoclonal
Clone: EPR27468-62
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat