Anti-Glucose Transporter GLUT3+GLUT14 Rabbit Monoclonal Antibody [clone: EPR10508(N)]

Lieferant: Abcam
AB191071-100 AB191071-1ML
ABCAAB191071-100EA 657 EUR
ABCAAB191071-100 ABCAAB191071-1ML
Anti-Glucose Transporter GLUT3+GLUT14 Rabbit Monoclonal Antibody [clone: EPR10508(N)]
Antikörper
Rabbit monoclonal [EPR10508(N)] to Glucose Transporter GLUT3 + GLUT14 - N-terminal.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glucose Transporter GLUT3+GLUT14
Clonality: Monoclonal
Clone: EPR10508(N)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR