Anti-GRAF Rabbit Monoclonal Antibody [clone: EPR8252]

Lieferant: Abcam
AB248808-100 AB248808-1
ABCAAB248808-100EA 1020 EUR
ABCAAB248808-100 ABCAAB248808-1
Anti-GRAF Rabbit Monoclonal Antibody [clone: EPR8252]
Antikörper
Rabbit monoclonal [EPR8252] to GRAF - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GRAF
Clonality: Monoclonal
Clone: EPR8252
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR