Anti-HLA-DR Rabbit Monoclonal Antibody [clone: EPR3692]

Lieferant: Abcam
AB215985-100 AB215985-1
ABCAAB215985-100EA 1020 EUR
ABCAAB215985-100 ABCAAB215985-1
Anti-HLA-DR Rabbit Monoclonal Antibody [clone: EPR3692]
Antikörper
Rabbit monoclonal [EPR3692] to HLA-DR - Low endotoxin, Azide free.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: EPR3692
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR