Anti-TLS/FUS Rat Monoclonal Antibody [clone: 9G6]

Lieferant: Abcam
AB255786-1 AB255786-100
ABCAAB255786-1EA 9160 EUR
ABCAAB255786-1 ABCAAB255786-100
Anti-TLS/FUS Rat Monoclonal Antibody [clone: 9G6]
Antikörper
Rat monoclonal [9G6] to TLS/FUS (methyl ) - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TLS/FUS
Clonality: Monoclonal
Clone: 9G6
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR