Anti-TLS/FUS Mouse Monoclonal Antibody [clone: Z-FUS-5]

Lieferant: Abcam
AB283272-100 AB283272-20UL AB283272-1ML
ABCAAB283272-100EA 782 EUR
ABCAAB283272-100 ABCAAB283272-20UL ABCAAB283272-1ML
Anti-TLS/FUS Mouse Monoclonal Antibody [clone: Z-FUS-5]
Antikörper
Mouse monoclonal [Z-FUS-5] to TLS/FUS - Mouse IgG2a.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TLS/FUS
Clonality: Monoclonal
Clone: Z-FUS-5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR