Anti-Glucose Transporter GLUT2 Rabbit Monoclonal Antibody [clone: EPR22946-74]

Lieferant: Abcam
AB234440-100 AB234440-20UL AB234440-1ML
ABCAAB234440-100EA 713 EUR
ABCAAB234440-100 ABCAAB234440-20UL ABCAAB234440-1ML
Anti-Glucose Transporter GLUT2 Rabbit Monoclonal Antibody [clone: EPR22946-74]
Antikörper
Rabbit monoclonal [EPR22946-74] to Glucose Transporter GLUT2.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glucose Transporter GLUT2
Clonality: Monoclonal
Clone: EPR22946-74
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR