Anti-DNA polymerase mu Rabbit Monoclonal Antibody [clone: EPR10470(B)]
Lieferant: Abcam
AB157465-100 AB157465-1ML
ABCAAB157465-100EA 607 EUR
ABCAAB157465-100 ABCAAB157465-1ML
Anti-DNA polymerase mu Rabbit Monoclonal Antibody [clone: EPR10470(B)]
Rabbit monoclonal [EPR10470(B)] to DNA polymerase mu.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: DNA polymerase mu
Clonality: Monoclonal
Clone: EPR10470(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human