Anti-Polyethylene glycol Rabbit Monoclonal Antibody [clone: EPR21993-154]

Lieferant: Abcam
AB203857-100 AB203857-1ML
ABCAAB203857-100EA 725 EUR
ABCAAB203857-100 ABCAAB203857-1ML
Anti-Polyethylene glycol Rabbit Monoclonal Antibody [clone: EPR21993-154]
Antikörper
Rabbit monoclonal [EPR21993-154] to Polyethylene glycol.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Polyethylene glycol
Clonality: Monoclonal
Clone: EPR21993-154
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR