Anti-cAMP Rabbit Monoclonal Antibody [clone: EP8471]

Lieferant: Abcam
AB179459-100 AB179459-1
ABCAAB179459-100EA 1020 EUR
ABCAAB179459-100 ABCAAB179459-1
Anti-cAMP Rabbit Monoclonal Antibody [clone: EP8471]
Antikörper
Rabbit monoclonal [EP8471] to cAMP - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: cAMP
Clonality: Monoclonal
Clone: EP8471
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR