Anti-Glucose Transporter 8 Rabbit Monoclonal Antibody [clone: EPR9477]

Lieferant: Abcam
AB169779-100 AB169779-1ML
ABCAAB169779-100EA 719 EUR
ABCAAB169779-100 ABCAAB169779-1ML
Anti-Glucose Transporter 8 Rabbit Monoclonal Antibody [clone: EPR9477]
Antikörper
Rabbit monoclonal [EPR9477] to Glucose Transporter 8.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glucose Transporter 8
Clonality: Monoclonal
Clone: EPR9477
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR