Anti-PINCH 1+PINCH-2 Rabbit Monoclonal Antibody [clone: EP1943Y]

Lieferant: Abcam
AB284740-100 AB284740-1
ABCAAB284740-100EA 1020 EUR
ABCAAB284740-100 ABCAAB284740-1
Anti-PINCH 1+PINCH-2 Rabbit Monoclonal Antibody [clone: EP1943Y]
Antikörper
Rabbit monoclonal [EP1943Y] to PINCH 1 + PINCH 2 - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PINCH 1+PINCH-2
Clonality: Monoclonal
Clone: EP1943Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR