Anti-LINE-1 ORF1p Rabbit Monoclonal Antibody [clone: EPR21844-108]

Lieferant: Abcam
AB216324-100 AB216324-1ML
ABCAAB216324-100EA 725 EUR
ABCAAB216324-100 ABCAAB216324-1ML
Anti-LINE-1 ORF1p Rabbit Monoclonal Antibody [clone: EPR21844-108]
Antikörper
Rabbit monoclonal [EPR21844-108] to LINE-1 ORF1p.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LINE-1 ORF1p
Clonality: Monoclonal
Clone: EPR21844-108
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR