N-(Chloracetyl)morpholin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

B21548.06 B21548.03
B21548.06EA 109 EUR
B21548.06 B21548.03
N-(Chloracetyl)morpholin ≥97%
N-(Chloracetyl)morpholin

Purity: 97+% CAS No.: 1440-61-5 Molecular Formula: C6H10ClNO2 Formula Weight: 163.61 Melting Point: 27-30° UN No.: 3261 MDL No.: MFCD00721939

Formel: C₆H₁₀ClNO₂
Schmelzpunkt: 27…30 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00721939
CAS-Nummer: 1440-61-5
EINECS: 215-880-3
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR