4-Phenylpyridin-N-oxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Phenylpyridin-1-ium-1-olate, 4-Phenylpyridine 1-oxide
M-Clarity™ Qualitätsstufe = MQ200
183490-25G
183490-5G
SIAL183490-25GEA
526
EUR
SIAL183490-25G
SIAL183490-5G
4-Phenylpyridin-N-oxid, Sigma-Aldrich®
4-Phenylpyridin-N-oxid
Formel:
C₁₁H₉NO Molekulargewicht: 171.1953 g/mol |
MDL Number:
MFCD00006208 CAS-Nummer: 1131-61-9 EINECS: 214-467-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...