Nickel(II)hydroxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
283622-1KG
283622-250G
SIAL283622-1KGEA
241
EUR
SIAL283622-1KG
SIAL283622-250G
Nickel(II)hydroxid, Sigma-Aldrich®
Nickel(II) hydroxide
Formel:
H₂NiO₂ |
MDL Number:
MFCD00011140 CAS-Nummer: 12054-48-7 EINECS: 235-008-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...