N-Fmoc-L-leucin

Lieferant: Cayman Chemical

10004888-25 10004888-50 10004888-100
CAYM10004888-25EA 27 EUR
CAYM10004888-25 CAYM10004888-50 CAYM10004888-100
N-Fmoc-L-leucin
N-Fmoc-L-leucin
Formel: C₂₁H₂₃NO₄
Molekulargewicht: 353.4 g/mol
MDL Number: MFCD00037133
CAS-Nummer: 35661-60-0
EINECS: 252-662-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR