Methylcyclopropancarboxylat
Lieferant: Thermo Scientific
Synonyme:
Methylcyclopropanecarboxylate
SB01039.1
SB01039.10
SB01039.50
MAYBSB01039.1EA
0
EUR
MAYBSB01039.1
MAYBSB01039.10
MAYBSB01039.50
Methylcyclopropancarboxylat
Methylcyclopropancarboxylat
Formel:
C₅H₈O₂ |
MDL Number:
MFCD00001280 CAS-Nummer: 2868-37-3 EINECS: 220-690-9 UN-Nummer: 3272 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...