2,4-Dimethylpyrrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dimethyl-1H-pyrrole

M-Clarity™ quality level = MQ200

390836-1G 390836-5G
SIAL390836-1GEA 50.4 EUR
SIAL390836-1G SIAL390836-5G
2,4-Dimethylpyrrol, Sigma-Aldrich®
2,4-Dimethylpyrrole
Formel: C₆H₉N
MDL Number: MFCD00192088
CAS-Nummer: 625-82-1
EINECS: 210-912-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR