3-Fluor-DL-phenylalanin ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH
L07297.03
L07297.06
L07297.03EA
49.5
EUR
L07297.03
L07297.06
3-Fluor-DL-phenylalanin ≥98%
3-Fluor-DL-phenylalanin
Purity: 98%
CAS No.: 2629-54-1
Molecular Formula: C9H10FNO2
Formula Weight: 183.18
Melting Point: ca 245° dec.
Beilstein Reference: 2939807
Formel:
C₉H₁₀FNO₂ Schmelzpunkt: ∼ 245 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004273 CAS-Nummer: 2629-54-1 EINECS: 220-104-1 UN-Nummer: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...