Borwasserstoff-Dimethylamin-Komplex, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Boron hydride dimethylamine, Borane, dimethylamine complex

M-Clarity™ quality level = MQ200

180238-100G 180238-25G 180238-5G
SIAL180238-100GEA 231 EUR
SIAL180238-100G SIAL180238-25G SIAL180238-5G
Borwasserstoff-Dimethylamin-Komplex, Sigma-Aldrich®
Borwasserstoff-Dimethylamin-Komplex
Formel: C₂H₁₀BN
MDL Number: MFCD00051068
CAS-Nummer: 74-94-2
EINECS: 200-823-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR