8-Iod-1-naphthoesäure
Lieferant: Thermo Scientific
NRB00572.250
NRB00572.1
MAYBNRB00572.250EA
0
EUR
MAYBNRB00572.250
MAYBNRB00572.1
8-Iod-1-naphthoesäure
8-Iod-1-naphthoesäure
Formel:
C₁₁H₇IO₂ |
CAS-Nummer:
13577-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...