Methyl-6-brom-2-naphthoat 98%
Lieferant: Thermo Fisher Scientific
364140050
364140250
364141000
ACRO364140050EA
36.1
EUR
ACRO364140050
ACRO364140250
ACRO364141000
Methyl-6-brom-2-naphthoat 98%
Methyl-6-brom-2-naphthoat
Formel:
C₁₂H₉BrO₂ Schmelzpunkt: 123…126 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00100408 CAS-Nummer: 33626-98-1 |
Spezifikation Testergebnisse
Appearance | Off-white to cream to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 118 to 126 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...