Diethyl(4-methoxybenzyl)phosphonat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Diethyl 4-methoxybenzylphosphonate
L05226.09
L05226.18
L05226.09EA
75.1
EUR
L05226.09
L05226.18
Diethyl(4-methoxybenzyl)phosphonat ≥98%
Diethyl(4-methoxybenzyl)phosphonat
Purity: 98+%
CAS No.: 1145-93-3
Molecular Formula: C12H19O4P
Formula Weight: 258.25
Flash Point:
Formel:
C₁₂H₁₉O₄P Siedepunkt: 136…138 °C (0.7 mmHg) Dichte: 1.500 Flammpunkt: >110 °C (230 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00015662 CAS-Nummer: 1145-93-3 EINECS: 214-548-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...