trans-2-Methoxy-β-nitrostyrol
Lieferant: Thermo Scientific
S11753.1
S11753.10
MAYBS11753.1EA
0
EUR
MAYBS11753.1
MAYBS11753.10
trans-2-Methoxy-β-nitrostyrol
trans-2-Methoxy-β-nitrostyrol
Formel:
C₉H₉NO₃ |
MDL Number:
MFCD00070647 CAS-Nummer: 3316-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...