Kaliumtetraoxalat Dihydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A14933.30
A14933.0I
A14933.0B
A14933.30EA
40.8
EUR
A14933.30
A14933.0I
A14933.0B
Kaliumtetraoxalat Dihydrat ≥98%
Kaliumtetraoxalat Dihydrat
Purity: 98+%
CAS No.: 6100-20-5
Molecular Formula: KH3(C2O4)2.2H2O
Formula Weight: 254.19 (218.16anhy)
Merck Reference: 14,7689
UN No.: 3288
MDL No.: MFCD00150443
Formel:
C₄H₃KO₈·2H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150443 CAS-Nummer: 6100-20-5 EINECS: 204-874-6 UN-Nummer: 3288 ADR: 6.1,III Merck Index: 14,7689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...