Dimethyl(2-oxopropyl)phosphonat ≥97%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Dimethyl acetylmethylphosphonate
L00537.06
L00537.14
L00537.06EA
103
EUR
L00537.06
L00537.14
Dimethyl(2-oxopropyl)phosphonat ≥97%
Dimethyl(2-oxopropyl)phosphonat
Purity: 97%
CAS No.: 4202-14-6
Molecular Formula: C5H11O4P
Formula Weight: 166.12
Flash Point: 93°(199°F)
Refractive Index: 1.4390
Boiling Point: 121-123°/11mm
Beilstein Reference: 1766218
MDL No.: MFCD00008769
Formel:
C₅H₁₁O₄P Siedepunkt: 121…123 °C (11 mmHg) Dichte: 1.202 Flammpunkt: 93 °C (199 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008769 CAS-Nummer: 4202-14-6 EINECS: 224-110-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...