Diethyl(2-bromethyl)phosphonat, Sigma-Aldrich®

Lieferant: Merck

D91152-25G D91152-5G
SIALD91152-25GEA 131 EUR
SIALD91152-25G SIALD91152-5G
Diethyl(2-bromethyl)phosphonat, Sigma-Aldrich®
Diethyl(2-bromethyl)phosphonat
Formel: C₆H₁₄BrO₃P
MDL Number: MFCD00000239
CAS-Nummer: 5324-30-1
EINECS: 226-193-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR