Ethyl-4-piperidincarboxylat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl piperidine-4-carboxylate, Ethyl isonipecotate, Ethyl 4-Piperidinecarboxylate
B20601.14
B20601.22
B20601.14EA
43.3
EUR
B20601.14
B20601.22
Ethyl-4-piperidincarboxylat ≥98%
Ethyl-4-piperidincarboxylat
Purity: 98%
CAS No.: 1126-09-6
Molecular Formula: C8H15NO2
Formula Weight: 157.21
Flash Point: 80°(176°F)
Refractive Index: 1.4600
Boiling Point: 203-205°
Beilstein Reference: 118419
MDL No.: MFCD00006003
Formel:
C₈H₁₅NO₂ Siedepunkt: 203…205 °C Dichte: 1.021 Flammpunkt: 80 °C (176 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00006003 CAS-Nummer: 1126-09-6 EINECS: 214-416-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...