5-Brom-1,3-benzodioxol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

M-Clarity™ quality level = MQ100

288314-100G 288314-25G
SIAL288314-100GEA 133 EUR
SIAL288314-100G SIAL288314-25G
5-Brom-1,3-benzodioxol, Sigma-Aldrich®
5-Brom-1,3-benzodioxol
Formel: C₇H₅BrO₂
MDL Number: MFCD00005821
CAS-Nummer: 2635-13-4
EINECS: 220-123-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR