5-Brom-1,3-benzodioxol

Lieferant: Thermo Scientific

Synonyme: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

CC01610.1 CC01610.10
MAYBCC01610.1EA 0 EUR
MAYBCC01610.1 MAYBCC01610.10
5-Brom-1,3-benzodioxol
5-Brom-1,3-benzodioxol
Formel: C₇H₅BrO₂
MDL Number: MFCD00005821
CAS-Nummer: 2635-13-4
EINECS: 220-123-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR