4,5,7-Trichlor-2-(trifluormethyl)chinolin

Lieferant: Thermo Scientific

KM07465.1 KM07465.10
MAYBKM07465.1EA 0 EUR
MAYBKM07465.1 MAYBKM07465.10
4,5,7-Trichlor-2-(trifluormethyl)chinolin
4,5,7-Trichloro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₃Cl₃F₃N
MDL Number: MFCD00153279
CAS-Nummer: 175203-41-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR