Bortrifluorid-Tetrahydrofuran-Komplex, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
434280-100ML
434280-500ML
SIAL434280-100MLEA
43.4
EUR
SIAL434280-100ML
SIAL434280-500ML
Bortrifluorid-Tetrahydrofuran-Komplex, Sigma-Aldrich®
Bortrifluorid-Tetrahydrofuran-Komplex
Formel:
C₄H₈BF₃O |
MDL Number:
MFCD00040372 CAS-Nummer: 462-34-0 EINECS: 207-325-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...