4-Amino-3-(trifluormethoxy)benzoesäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

H32915.06 H32915.14
H32915.06EA 99.1 EUR
H32915.06 H32915.14
4-Amino-3-(trifluormethoxy)benzoesäure 98%
4-Amino-3-(trifluoromethoxy)benzoic acid

Purity: 98% CAS No.: 175278-22-5 Molecular Formula: C8H6F3NO3 Formula Weight: 221.14 MDL No.: MFCD00205143

Formel: C₈H₆F₃NO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00205143
CAS-Nummer: 175278-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR