4-(2-Pyridylazo)resorcin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L09667.06
L09667.03
L09667.06EA
92.1
EUR
L09667.06
L09667.03
4-(2-Pyridylazo)resorcin ≥99%
4-(2-Pyridylazo)resorcin
Purity: 99%
CAS No.: 1141-59-9
Molecular Formula: C11H9N3O2
Formula Weight: 215.21
Melting Point: ca 187° dec.
Beilstein Reference: 184665
Formel:
C₁₁H₉N₃O₂ Schmelzpunkt: ∼ 187 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00044237 CAS-Nummer: 1141-59-9 EINECS: 214-528-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...