4-(2-Pyridylazo)resorcin ACS

Lieferant: Thermo Fisher Scientific
Achtung

43629.03 43629.06
43629.03EA 41.9 EUR
43629.03 43629.06
4-(2-Pyridylazo)resorcin ACS
4-(2-Pyridylazo)resorcin

CAS No.: 1141-59-9 Molecular Formula: C11H9N3O2 Formula Weight: 215.21 Physical Form: Powder Melting Point: 187° dec. Beilstein Reference: 184665

Formel: C₁₁H₉N₃O₂
Schmelzpunkt: 187 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00044237
CAS-Nummer: 1141-59-9
EINECS: 214-528-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR