4-(4-Nitrophenylazo)resorcin ≥90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-((4-Nitrophenyl)diazenyl)benzene-1,3-diol, Azoviolet, Azo Violet, Magneson I

A12232.14 A12232.22 A12232.36
A12232.14EA 47.3 EUR
A12232.14 A12232.22 A12232.36
4-(4-Nitrophenylazo)resorcin ≥90%
4-(4-Nitrophenylazo)resorcin

Purity: 95% CAS No.: 74-39-5 Molecular Formula: C12H9N3O4 Formula Weight: 259.22 Melting Point: ca 185° dec. Beilstein Reference: 674709 Merck Reference: 14,5695 MDL No.: MFCD00007310

Formel: C₁₂H₉N₃O₄
Molekulargewicht: 259.22 g/mol
Schmelzpunkt: ∼ 185 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007310
CAS-Nummer: 74-39-5
EINECS: 200-808-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR