Diethyl(2-cyanethyl)malonat

Lieferant: Thermo Scientific

Synonyme: Diethyl 2-(2-cyanoethyl)malonate

SB01209.1 SB01209.10 SB01209.50
MAYBSB01209.1EA 0 EUR
MAYBSB01209.1 MAYBSB01209.10 MAYBSB01209.50
Diethyl(2-cyanethyl)malonat
Diethyl(2-cyanethyl)malonat
Formel: C₁₀H₁₅NO₄
MDL Number: MFCD00001966
CAS-Nummer: 17216-62-5
EINECS: 241-260-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR