N-Methyl-1,3-propandiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Methylpropane-1,3-diamine, 3-Methylamino propylamine
M-Clarity™ quality level = MQ200
127027-500G
127027-100G
127027-25G
SIAL127027-500GEA
185
EUR
SIAL127027-500G
SIAL127027-100G
SIAL127027-25G
N-Methyl-1,3-propandiamin, Sigma-Aldrich®
N-Methyl-1,3-propandiamin
Formel:
C₄H₁₂N₂ |
MDL Number:
MFCD00008209 CAS-Nummer: 6291-84-5 EINECS: 228-544-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...