Tetrabutylammoniumacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetra-n-butylammonium acetate

M-Clarity™ quality level = MQ200

335991-50G 335991-10G 335991-5G
SIAL335991-50GEA 428 EUR
SIAL335991-50G SIAL335991-10G SIAL335991-5G
Tetrabutylammoniumacetat, Sigma-Aldrich®
Tetrabutylammoniumacetat
Formel: C₁₈H₃₉NO₂
MDL Number: MFCD00043208
CAS-Nummer: 10534-59-5
EINECS: 234-101-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR