N-Acetyl-L-methionin ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH
B24312.06
B24312.14
B24312.06EA
53.7
EUR
B24312.06
B24312.14
N-Acetyl-L-methionin ≥99%
N-Acetyl-L-methionin
Purity: 99%
CAS No.: 65-82-7
Molecular Formula: C7H13NO3S
Formula Weight: 191.25
Melting Point: 103-107°
Beilstein Reference: 1725552
Merck Reference: 14,96
Optical Rotation: -21° (c=1.3 in water)
MDL No.: MFCD00064441
Formel:
C₇H₁₃NO₃S Schmelzpunkt: 103…107 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064441 CAS-Nummer: 65-82-7 EINECS: 200-617-7 Merck Index: 14,96 |