N-Acetyl-DL-methionin ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ac-DL-Met-OH
B21866.0B
B21866.18
B21866.30
B21866.0BEA
200
EUR
B21866.0B
B21866.18
B21866.30
N-Acetyl-DL-methionin ≥99%
N-Acetyl-DL-methionin
Purity: 99%
CAS No.: 1115-47-5
Molecular Formula: C7H13NO3S
Formula Weight: 191.25
Storage Temperature: 2°C to 8°C
Melting Point: 113-117°
Beilstein Reference: 1725554
Merck Reference: 14,96
MDL No.: MFCD00008681
Formel:
C₇H₁₃NO₃S Schmelzpunkt: 113…117 °C |
MDL Number:
MFCD00008681 CAS-Nummer: 1115-47-5 EINECS: 214-224-3 Merck Index: 14,96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...