2',7'-Dichlorfluorescein

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

A16212.06 A16212.14
A16212.06EA 65.5 EUR
A16212.06 A16212.14
2',7'-Dichlorfluorescein
2',7'-Dichlorfluorescein

CAS No.: 76-54-0 Molecular Formula: C20H10Cl2O5 Formula Weight: 401.20 Melting Point: 300° dec. MDL No.: MFCD00005047

Formel: C₂₀H₁₀Cl₂O₅
Schmelzpunkt: >300 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00005047
CAS-Nummer: 76-54-0
EINECS: 200-968-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR