4-(2-Pyridylazo)resorcin Mononatriumsalz Monohydrat, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: PAR monosodium salt monohydrate, 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, PAR

M-Clarity™ Qualitätsstufe = MQ200

178268-10G 178268-5G
178268-10GEA 280 EUR
178268-10G 178268-5G
4-(2-Pyridylazo)resorcin Mononatriumsalz Monohydrat, Sigma-Aldrich®
4-(2-Pyridylazo)resorcin Mononatriumsalz Monohydrat

4-(2-Pyridylazo)resorcinol monosodium salt is a chromophoric indicator and chromogenic chelator employed for the spectrophotometric determination of several metal ions.


4-(2-Pyridylazo)resorcinol monosodium salt hydrate has been used in the preparation of titanium reagent for the spectrometric measurement of hydrogen peroxide (H₂O₂) and superoxide anion (O₂•-). It is also suitable to measure the concentration of Zn²⁺, Zinc transition metal ions.

Formel: C₁₁H₈N₃NaO₂·1H₂O
Molekulargewicht: 255.21 g/mol (237.19 anhy)
Lagerungstemperatur: Room Temperature
MDL Number: MFCD00150712
CAS-Nummer: 16593-81-0
EINECS: 236-339-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c121e63e562bbcbe0b3ee93740cbe5ee6edb24a6&vid=5dfbf6928c7c58c2c0b7cf85643e45c47de43e7a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity (HPLC area %) ≥ 97.5 %
Water 4 - 10 %
Infrared Spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR