Tetrabutylammoniumdihydrogenphosphat 1 mol/l in wässriger Lösung, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate
M-Clarity™ quality level = MQ200
268100-100ML
268100-500ML
SIAL268100-100MLEA
130
EUR
SIAL268100-100ML
SIAL268100-500ML
Tetrabutylammoniumdihydrogenphosphat 1 mol/l in wässriger Lösung, Sigma-Aldrich®
Tetrabutylammoniumdihydrogenphosphat
Formel:
C₁₆H₃₈NO₄P |
MDL Number:
MFCD00064526 CAS-Nummer: 5574-97-0 EINECS: 226-947-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...