Ethyl 3-isothiocyanatopropionat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Isothiocyanatopropionic acid ethyl ester

L10911.03 L10911.06
L10911.03EA 72.8 EUR
L10911.03 L10911.06
Ethyl 3-isothiocyanatopropionat ≥97%
Ethyl 3-isothiocyanatopropionat

Purity: 97% CAS No.: 17126-62-4 Molecular Formula: C6H9NO2S Formula Weight: 159.20 Refractive Index: 1.5014 Odour: Pungent Boiling Point: 75-76°/8mm Beilstein Reference: 1705633 UN No.: 2922 MDL No.: MFCD00041130

Formel: C₆H₉NO₂S
Siedepunkt: 75…76 °C (8 mmHg)
Dichte: 1.12
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00041130
CAS-Nummer: 17126-62-4
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR