Bis(pentamethylcyclopentadienyl)eisen(II), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
378542-1G
378542-5G
SIAL378542-1GEA
181
EUR
SIAL378542-1G
SIAL378542-5G
Bis(pentamethylcyclopentadienyl)eisen(II), Sigma-Aldrich®
Bis(pentamethylcyclopentadienyl)eisen(II)
Formel:
C₂₀H₃₀Fe |
MDL Number:
MFCD00058736 CAS-Nummer: 12126-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...