Calcein, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

C0875-5G C0875-25G
SIALC0875-5GEA 142 EUR
SIALC0875-5G SIALC0875-25G
Calcein, Sigma-Aldrich®
Calcein
Formel: C₃₀H₂₆N₂O₁₃
MDL Number: MFCD00005049
CAS-Nummer: 1461-15-0
EINECS: 215-957-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR